Di-tert-dodecyl disulfide (mixture of isomers)

Supplier: TCI America

D0447-25G D0447-250G
TCD0447-025GEA 105.86 CAD
TCD0447-025G TCD0447-250G
Di-tert-dodecyl disulfide (mixture of isomers)
Di-tert-dodecyl disulfide (mixture of isomers)

CAS Number: 27458-90-8
MDL Number: MFCD00059944
Molecular Formula: C24H50S2
Molecular Weight: 402.78
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 321
Flash Point (°C): 118
Specific Gravity (20/20): 0.91


Other Product Names / Synonyms: tert-Dodecyl Disulfide

Formula: C₂₄H₅₀S₂
Boiling Pt: 321 °C
Density: 0.91 (20 °C)
Flash Pt: 118 °C
MDL Number: MFCD00059944
CAS Number: 27458-90-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4bb0095552e420ce68ab5244c3b9b091dbb0e3da&vid=4bb0095552e420ce68ab5244c3b9b091dbb0e3da&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR