Guanylurea sulfate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

D0433-25G D0433-500G
TCD0433-025GEA 83.74 CAD
TCD0433-025G TCD0433-500G
Guanylurea sulfate ≥98.0% (by titrimetric analysis)
Guanylurea sulfate

CAS Number: 591-01-5
MDL Number: MFCD00035565
Molecular Formula: C2H6N4O
Molecular Weight: 302.27
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 190


Other Product Names / Synonyms: Dicyandiamidine Sulfate

Formula: C₂H₈N₄O₅S
Melting Pt: 190 °C
MDL Number: MFCD00035565
CAS Number: 591-01-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bbbe73957dcffb0e20a8ba013b2c07ea84b6d7d7&vid=bbbe73957dcffb0e20a8ba013b2c07ea84b6d7d7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR