2',7'-Dichlorofluorescein disodium salt ≥98.0%
Supplier: TCI America
Synonyms:
Sodium 2',7'-dichloro-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-bis(olate), 2',7'-Dichlorofluorescein sodium salt
D0424-25G
D0424-1G
TCD0424-025GEA
647.8
CAD
TCD0424-025G
TCD0424-001G
2',7'-Dichlorofluorescein disodium salt ≥98.0%
2',7'-Dichlorofluorescein disodium salt
CAS Number: 80471-69-8
MDL Number: MFCD00070511
Molecular Formula: C20H10Cl2O5
Molecular Weight: 445.16
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Formula:
C₂₀H₈Cl₂Na₂O₅ MW: 445.160 g/mol |
MDL Number:
MFCD00070511 CAS Number: 80471-69-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...