2-Amino-4-chloro-5-methylbenzenesulfonic acid sodium salt ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: 5-Chloro-4-methylaniline-2-sulphonic acid Sodium Salt, Sodium 2-amino-4-chloro-5-methylbenzenesulfonate, Sodium 2-Amino-4-chloro-5-methylbenzenesulfonate, 5-Chloro-4-methylaniline-2-sulfonic acid Sodium Salt, Aminochlorotoluenesulfonicacidsodiumsalt, Sodium 2-amino-4-chloro-5-methylbenzenesulphonate, Aminochlorotoluenesulphonicacidsodiumsalt, 5-Chloro-p-toluidine-2-sulphonic acid Sodium Salt, Sodium 2-Amino-4-chloro-5-methylbenzenesulphonate, 5-Chloro-p-toluidine-2-sulfonic acid Sodium Salt

A0896-25G A0896-500G
TCA0896-025GEA 26.86 CAD
TCA0896-025G TCA0896-500G
2-Amino-4-chloro-5-methylbenzenesulfonic acid sodium salt ≥98.0% (by HPLC, total nitrogen)
2-Amino-4-chloro-5-methylbenzenesulphonic acid sodium salt

CAS Number: 6627-59-4
MDL Number: MFCD00070551
Molecular Formula: C7H8ClNO3S
Molecular Weight: 243.64
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal

Formula: C₇H₇ClNNaO₃S
MW: 243.6432 g/mol
MDL Number: MFCD00070551
CAS Number: 6627-59-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6820ec1bbc8a080b42d6b70a54ec846e44fc4005&vid=6820ec1bbc8a080b42d6b70a54ec846e44fc4005&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR