2,5-Dichlorosulfanilic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

D0415-500G D0415-25G
TCD0415-500GEA 2543.8 CAD
TCD0415-500G TCD0415-025G
2,5-Dichlorosulfanilic acid ≥98.0% (by HPLC, titration analysis)
2,5-Dichlorosulfanilic Acid

CAS Number: 88-50-6
MDL Number: MFCD00025159
Molecular Formula: C6H5Cl2NO3S
Molecular Weight: 242.07
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal


Other Product Names / Synonyms: 2,5-Dichloroaniline-4-sulfonic Acid

Formula: C₆H₅Cl₂NO₃S
MDL Number: MFCD00025159
CAS Number: 88-50-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=403a3447a5ec5bbc7098d90cc2286da3006bfa73&vid=403a3447a5ec5bbc7098d90cc2286da3006bfa73&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR