Fluorescein chloride ≥95.0% (by HPLC)

Supplier: TCI America

D0369-1G D0369-25G
TCD0369-001GEA 41.08 CAD
TCD0369-001G TCD0369-025G
Fluorescein chloride ≥95.0% (by HPLC)
Fluorescein chloride

[Reagent for Amines]
CAS Number: 630-88-6
MDL Number: MFCD00037841
Molecular Formula: C20H10Cl2O3
Molecular Weight: 369.20
Purity/Analysis Method: >95.0% (HPLC)
Form: Crystal


Product Categories: Analytical Science, Fluorescent Dyes
Other Product Names / Synonyms: 3',6'-Dichlorofluoran

Formula: C₂₀H₁₀Cl₂O₃
MDL Number: MFCD00037841
CAS Number: 630-88-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=90adcc2a0c19eb15a1b34622f6098a9d0fa23ec0&vid=90adcc2a0c19eb15a1b34622f6098a9d0fa23ec0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR