Methyl-DL-2-aminobutyrate hydrochloride ≥98.0%

Supplier: TCI America

Synonyms: H-Abu-OMe·HCl, DL-2-Aminobutyric Acid Methyl Ester Hydrochloride, Methyl 2-aminobutanoate hydrochloride

A0893-25G A0893-5G
TCA0893-025GEA 557.74 CAD
TCA0893-025G TCA0893-5G
Methyl-DL-2-aminobutyrate hydrochloride ≥98.0%
Methyl-DL-2-aminobutyrate hydrochloride

CAS Number: 7682-18-0
MDL Number: MFCD00058295
Molecular Formula: C5H11NO2
Molecular Weight: 153.61
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 150


Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: DL-2-Aminobutyric Acid Methyl Ester Hydrochloride

Formula: C₅H₁₁NO₂
MW: 153.61 g/mol
Melting Pt: 150 °C
MDL Number: MFCD00058295
CAS Number: 7682-18-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f21de271a96bd13d2d7d74457c3c7862a2e59389&vid=f21de271a96bd13d2d7d74457c3c7862a2e59389&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR