2,5-Di-tert-amylhydroquinone ≥93.0%

Supplier: TCI America

Synonyms: 2,5-Di-tert-pentylhydroquinone

D0299-25G D0299-500G
TCD0299-025GEA 57.84 CAD
TCD0299-025G TCD0299-500G
2,5-Di-tert-amylhydroquinone ≥93.0%
2,5-Di-tert-amylhydroquinone

CAS Number: 79-74-3
MDL Number: MFCD00027029
Molecular Formula: C16H26O2
Molecular Weight: 250.38
Purity/Analysis Method: >93.0% (GC)
Form: Crystal
Melting point (°C): 179


Product Categories: Materials Chemistry, Resin Additives
Other Product Names / Synonyms: 2,5-Di-tert-pentylhydroquinone

Formula: C₁₆H₂₆O₂
MW: 250.38 g/mol
Melting Pt: 179 °C
MDL Number: MFCD00027029
CAS Number: 79-74-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5131c53417af94ca5fbac7d838741c0ebd1888e2&vid=5131c53417af94ca5fbac7d838741c0ebd1888e2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 93.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR