2-[2-(Diethylamino)ethoxy]ethanol ≥98.0%

Supplier: TCI America

Synonyms: Ethylene Glycol Mono[2-(diethylamino)ethyl] Ether, 2-(2-(Diethylamino)ethoxy)ethanol

D0298-25G D0298-500G
TCD0298-025GEA 50.56 CAD
TCD0298-025G TCD0298-500G
2-[2-(Diethylamino)ethoxy]ethanol ≥98.0%
2-[2-(Diethylamino)ethoxy]ethanol

CAS Number: 140-82-9
MDL Number: MFCD00020604
Molecular Formula: C8H19NO2
Molecular Weight: 161.25
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 101
Flash Point (°C): 96
Specific Gravity (20/20): 0.94


Other Product Names / Synonyms: Ethylene Glycol Mono[2-(diethylamino)ethyl] Ether

Formula: C₈H₁₉NO₂
MW: 161.25 g/mol
Boiling Pt: 101 °C
Density: 0.94 (20 °C)
Flash Pt: 96 °C
MDL Number: MFCD00020604
CAS Number: 140-82-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=42bf2dfc8cdab31982f1ae0aa18c1f85a4d695c4&vid=42bf2dfc8cdab31982f1ae0aa18c1f85a4d695c4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR