4,4'-Dithiodimorpholine ≥98.0%
Supplier: TCI America
Synonyms:
N,N'-Dimorpholine Disulfide, 4-(4-Morpholinyldisulfanyl)morpholine, N,N'-Dithiobis(morpholine), 4-(4-Morpholinyldisulphanyl)morpholine, N,N'-Dimorpholine Disulphide
CAS Number: 103-34-4
MDL Number: MFCD00023319
Molecular Formula: C8H16N2O2S2
Molecular Weight: 236.35
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 125
Flash Point (°C): 177
Storage Temperature: 0-10°C
Other Product Names / Synonyms: N,N'-Dimorpholine Disulfide, N,N'-Dithiobis(morpholine)
Formula:
C₈H₁₆N₂O₂S₂ Melting Pt: 125 °C Flash Pt: 177 °C |
MDL Number:
MFCD00023319 CAS Number: 103-34-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...