2,4-Dinitro-N-phenylaniline ≥98.0%

Supplier: TCI America

Synonyms: 2,4-Dinitrodiphenylamine

D0278-25G D0278-100G
TCD0278-025GEA 105.86 CAD
TCD0278-025G TCD0278-100G
2,4-Dinitro-N-phenylaniline ≥98.0%
2,4-Dinitro-N-phenylaniline

CAS Number: 961-68-2
MDL Number: MFCD00007223
Molecular Formula: C12H9N3O4
Molecular Weight: 259.22
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 157

Formula: C₁₂H₉N₃O₄
MW: 259.22 g/mol
Melting Pt: 157 °C
MDL Number: MFCD00007223
CAS Number: 961-68-2
EINECS: 213-508-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9acb4424a616e8f76705e5f4d802340660ade683&vid=9acb4424a616e8f76705e5f4d802340660ade683&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR