4-Methoxybenzyl acetate ≥98.0%

Supplier: TCI America

A0886-25ML A0886-500ML
TCA0886-025MLEA 52.14 CAD
TCA0886-025ML TCA0886-500ML
4-Methoxybenzyl acetate ≥98.0%
4-Methoxybenzyl acetate

CAS Number: 104-21-2
MDL Number: MFCD00038509
Molecular Formula: C10H12O3
Molecular Weight: 180.20
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 137
Flash Point (°C): 140
Specific Gravity (20/20): 1.11


Other Product Names / Synonyms: Acetic Acid 4-Methoxybenzyl Ester, Anisyl Acetate, Acetic Acid Anisyl Ester

Formula: C₁₀H₁₂O₃
Boiling Pt: 137 °C
Density: 1.11 (20 °C)
Flash Pt: 140 °C
MDL Number: MFCD00038509
CAS Number: 104-21-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=52dc778da440596489c600136ab2d85f0e3218b0&vid=52dc778da440596489c600136ab2d85f0e3218b0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR