2-Benzothiazolyl diethyldithiocarbamate ≥95.0%

Supplier: TCI America

Synonyms: Benzo[d]thiazol-2-yl diethylcarbamodithioate, Diethyldithiocarbamic acid 2-Benzothiazolyl Ester, N,N-Diethylthiocarbamoyl-2-benzothiazolyl sulphide, N,N-Diethylthiocarbamoyl-2-benzothiazolyl Sulfide

D0248-500G D0248-25G
TCD0248-500GEA 379.2 CAD
TCD0248-500G TCD0248-025G
2-Benzothiazolyl diethyldithiocarbamate ≥95.0%
2-Benzothiazolyl diethyldithiocarbamate

CAS Number: 95-30-7
MDL Number: MFCD00059887
Molecular Formula: C12H14N2S3
Molecular Weight: 282.44
Purity/Analysis Method: >95.0% (N)
Form: Crystal
Melting point (°C): 79


Other Product Names / Synonyms: Diethyldithiocarbamic Acid 2-Benzothiazolyl Ester, N,N-Diethylthiocarbamoyl-2-benzothiazolyl Sulfide

Formula: C₁₂H₁₄N₂S₃
MW: 282.44 g/mol
Melting Pt: 79 °C
MDL Number: MFCD00059887
CAS Number: 95-30-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=243ff732e4970058c1b49ec501b12b83995b390d&vid=243ff732e4970058c1b49ec501b12b83995b390d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR