4,4'-Dibenzoylquinone dioxime ≥98.0%
Supplier: TCI America
D0246-25G
D0246-500G
TCD0246-025GEA
44.24
CAD
TCD0246-025G
TCD0246-500G
4,4'-Dibenzoylquinone dioxime ≥98.0%
4,4'-Dibenzoylquinone dioxime
CAS Number: 120-52-5
MDL Number: MFCD00045557
Molecular Formula: C20H14N2O4
Molecular Weight: 346.34
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Flash Point (°C): 160
Formula:
C₂₀H₁₄N₂O₄ MW: 346.34 g/mol Flash Pt: 160 °C |
MDL Number:
MFCD00045557 CAS Number: 120-52-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...