Dibutyl disulfide ≥95.0% for synthesis

Supplier: TCI America
Warning

Synonyms: Dibutyldisulphide, n-Butyl disulfide, n-Butyl disulphide, Dibutyldisulfide

D0224-25ML D0224-500ML
TCD0224-025MLEA 77.42 CAD
TCD0224-025ML TCD0224-500ML
Dibutyl disulfide ≥95.0% for synthesis
Dibutyl disulphide

CAS Number: 629-45-8
MDL Number: MFCD00009467
Molecular Formula: C8H18S2
Molecular Weight: 178.35
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 88
Flash Point (°C): 93
Specific Gravity (20/20): 0.94


Other Product Names / Synonyms: Butyl Disulfide

Formula: C₈H₁₈S₂
MW: 178.36 g/mol
Boiling Pt: 229
Melting Pt: -94
Density: 0.94 (20 °C)
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00009467
CAS Number: 629-45-8
EINECS: 211-091-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=50bd5f84aa43cacace3d8ba3a8fd653cb611cd62&vid=50bd5f84aa43cacace3d8ba3a8fd653cb611cd62&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR