Bromopyrogallol red

Supplier: TCI America

D0207-5G D0207-1G
TCD0207-5GEA 287.56 CAD
TCD0207-5G TCD0207-001G
Bromopyrogallol red
Bromopyrogallol red

CAS Number: 16574-43-9
MDL Number: MFCD00148865
Molecular Formula: C19H10Br2O8S
Molecular Weight: 558.15
Form: Crystal
Color: Yellow Red


Other Product Names / Synonyms: Dibromopyrogallolsulfonphthaleine

Formula: C₁₉H₁₀Br₂O₈S
MDL Number: MFCD00148865
CAS Number: 16574-43-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f87547bb2c68a61a5944d7e582f4aa49beaffa3a&vid=f87547bb2c68a61a5944d7e582f4aa49beaffa3a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR