Ethylene glycol dibenzyl ether ≥95.0%
Supplier: TCI America
D0160-25ML
D0160-5ML
TCD0160-025MLEA
377.62
CAD
TCD0160-025ML
TCD0160-5ML
Ethylene glycol dibenzyl ether ≥95.0%
Ethylene glycol dibenzyl ether
CAS Number: 622-22-0
MDL Number: MFCD00030016
Molecular Formula: C16H18O2
Molecular Weight: 242.32
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 210
Specific Gravity (20/20): 1.06
Other Product Names / Synonyms: 1,2-Bis(benzyloxy)ethane, Dibenzyl Cellosolve, Dibenzyl Glycol, 1,2-Dibenzyloxyethane
Formula:
C₁₆H₁₈O₂ Boiling Pt: 210 °C Density: 1.06 (20 °C) |
MDL Number:
MFCD00030016 CAS Number: 622-22-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...