N-Acetyl-L-histidine monohydrate ≥99.0%

Supplier: TCI America

Synonyms: Nα-Acetyl-L-histidine monohydrate, Nα-Acetyl-L-histidine, N-Acetyl-L-histidine Monohydrate, Ac-His-OH.H2O, Ac-His-OH·H2O, N-Ac-His-OH·H₂O

A0873-1G A0873-25G
TCA0873-001GEA 63.2 CAD
TCA0873-001G TCA0873-025G
N-Acetyl-L-histidine monohydrate ≥99.0%
N-Acetyl-L-histidine monohydrate

CAS Number: 39145-52-3
MDL Number: MFCD00149320
Molecular Formula: C8H11N3O3
Molecular Weight: 197.19
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 163
Specific rotation [a]20/D: 46.5 deg (C=1, H2O)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: Ac-His-OH

Formula: C₈H₁₁N₃O₃·H₂O
MW: 215.2 g/mol
Melting Pt: 163 °C
MDL Number: MFCD00149320
CAS Number: 39145-52-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c860dd1b11cf86b0acda40ba6bdaaec2f3fefcd7&vid=c860dd1b11cf86b0acda40ba6bdaaec2f3fefcd7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR