Sodium dibenzyldithiocarbamate hydrate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

D0156-25G D0156-5G
TCD0156-025GEA 611.46 CAD
TCD0156-025G TCD0156-005G
Sodium dibenzyldithiocarbamate hydrate ≥98.0% (by titrimetric analysis)
Sodium dibenzyldithiocarbamate hydrate

CAS Number: 55310-46-8
MDL Number: MFCD00066462
Molecular Formula: C15H15NS2
Molecular Weight: 295.39
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Other Product Names / Synonyms: Dibenzyldithiocarbamic Acid Sodium Salt

Formula: C₁₅H₁₅NS₂
MDL Number: MFCD00066462
CAS Number: 55310-46-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4b90b74306b66e1ca295172c0d2bba810bc93ed0&vid=b9df075251034b982fc763f0dfc36fb57a2dee63&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR