N,N-Dibenzyl-2-aminoethanol ≥98.0%

Supplier: TCI America

Synonyms: N-(2-Hydroxyethyl)dibenzylamine, N,N-Dibenzylethanolamine

D0153-25G D0153-500G
TCD0153-025GEA 167.24 CAD
TCD0153-025G TCD0153-500G
N,N-Dibenzyl-2-aminoethanol ≥98.0%
N,N-Dibenzyl-2-aminoethanol

CAS Number: 101-06-4
MDL Number: MFCD00020574
Molecular Formula: C16H19NO
Molecular Weight: 241.33
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Boiling point (°C): 206
Freezing point (°C): 38


Other Product Names / Synonyms: N,N-Dibenzylethanolamine, N-(2-Hydroxyethyl)dibenzylamine

Formula: C₁₆H₁₉NO
MW: 241.33 g/mol
Boiling Pt: 206 °C
MDL Number: MFCD00020574
CAS Number: 101-06-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea50ac2e434fd8d066329db89edaf6c06ec27aaf&vid=ea50ac2e434fd8d066329db89edaf6c06ec27aaf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR