4,4'-Diaminostilbene-2,2'-disulfonic Acid ≥94.0%
Supplier: TCI America
Synonyms:
Amsonic acid, 4,4-Diamino-2,2-stilbenedisulphonic acid, 4,4-Diamino-2,2-stilbenedisulfonic acid
D0121-25G
D0121-500G
D0121-100G
TCD0121-025GEA
60.04
CAD
TCD0121-025G
TCD0121-500G
TCD0121-100G
4,4'-Diaminostilbene-2,2'-disulfonic Acid ≥94.0%
4,4'-Diaminostilbene-2,2'-disulfonic Acid
CAS Number: 81-11-8
MDL Number: MFCD00024946
Molecular Formula: C14H14N2O6S2
Molecular Weight: 370.39
Purity/Analysis Method: >94.0% (T)
Form: Crystal
Product Categories: Materials Chemistry, Monomers, Polyimide Raw Materials
Other Product Names / Synonyms: Amsonic Acid
Formula:
C₁₄H₁₄N₂O₆S₂ MW: 370.39 g/mol |
MDL Number:
MFCD00024946 CAS Number: 81-11-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...