4,4'-Diaminostilbene dihydrochloride ≥98.0%

Supplier: TCI America

D0120-5G D0120-1G
TCD0120-005GEA 687.3 CAD
TCD0120-005G TCD0120-001G
4,4'-Diaminostilbene dihydrochloride ≥98.0%
4,4'-Diaminostilbene dihydrochloride

CAS Number: 54760-75-7
MDL Number: MFCD00012984
Molecular Formula: C14H14N2
Molecular Weight: 283.20
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal

Formula: C₁₄H₁₄N₂
MDL Number: MFCD00012984
CAS Number: 54760-75-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea685f6254e49babb20c19e499c5c778522b2ef0&vid=ea685f6254e49babb20c19e499c5c778522b2ef0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR