1,5-Diaminopentane dihydrochloride ≥98.0% (by total nitrogen basis)

Supplier: TCI America

D0099-25G D0099-5G
TCD0099-25GEA 418.7 CAD
TCD0099-25G TCD0099-005G
1,5-Diaminopentane dihydrochloride ≥98.0% (by total nitrogen basis)
1,5-Diaminopentane dihydrochloride

CAS Number: 1476-39-7
MDL Number: MFCD00012527
Molecular Formula: C5H14N2
Molecular Weight: 175.10
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 230


Product Categories: Materials Chemistry, Monomers, Bioscience, Polyamines
Other Product Names / Synonyms: Cadaverine Hydrochloride, Pentamethylenediamine Hydrochloride, 1,5-Pentanediamine Dihydrochloride

Formula: NH2(CH2)5NH2 .2HCl
Melting Pt: 230 °C
MDL Number: MFCD00012527
CAS Number: 1476-39-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=05df70ab93310cbf448343a9a6d6b78c16087be9&vid=05df70ab93310cbf448343a9a6d6b78c16087be9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR