trans-2-Decenoic acid ≥95.0% (by titrimetric analysis)

Supplier: TCI America

D0098-25ML D0098-5ML
TCD0098-25MLEA 302.16 CAD
TCD0098-25ML TCD0098-5ML
trans-2-Decenoic acid ≥95.0% (by titrimetric analysis)
trans-2-Decenoic acid

CAS Number: 334-49-6
MDL Number: MFCD00039530
Molecular Formula: C10H18O2
Molecular Weight: 170.25
Purity/Analysis Method: >95.0% (T)
Form: Clear Liquid
Boiling point (°C): 121
Flash Point (°C): 170
Specific Gravity (20/20): 0.93


Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: trans-2-Decylenic Acid

Formula: C₁₀H₁₈O₂
Boiling Pt: 121 °C
Density: 0.93 (20 °C)
Flash Pt: 170 °C
MDL Number: MFCD00039530
CAS Number: 334-49-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a87233f06d4448f089fc36ae1d72a3c0770773e2&vid=a87233f06d4448f089fc36ae1d72a3c0770773e2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR