N(α)-Acetyl-D-tryptophan ≥98.0%

Supplier: TCI America

Synonyms: H-D-Trp-OH, Ac-D-Trp-OH, N-Acetyl-D-tryptophan

A0119-5G A0119-1G
TCA0119-005GEA 123.24 CAD
TCA0119-005G TCA0119-001G
N(α)-Acetyl-D-tryptophan ≥98.0%
N(α)-Acetyl-D-tryptophan

CAS Number: 2280-01-5
MDL Number: MFCD00065021
Molecular Formula: C13H14N2O3
Molecular Weight: 246.27
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 186


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: Ac-D-Trp-OH

Formula: C₁₃H₁₄N₂O₃
MW: 246.3 g/mol
Melting Pt: 186 °C
MDL Number: MFCD00065021
CAS Number: 2280-01-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3547f67049143cfcbf20be78cf2b3ccbadb678df&vid=3547f67049143cfcbf20be78cf2b3ccbadb678df&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR