Sodium decanoate ≥99.0% (by titrimetric analysis)
Supplier: TCI America
D0024-25G
D0024-500G
D0024-100G
TCD0024-025GEA
136.91
CAD
TCD0024-025G
TCD0024-500G
TCD0024-100G
Sodium decanoate ≥99.0% (by titrimetric analysis)
Sodium decanoate
CAS Number: 1002-62-6
MDL Number: MFCD00066453
Molecular Formula: C10H20O2
Molecular Weight: 194.25
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Decanoic Acid Sodium Salt
Formula:
C₁₀H₁₉NaO₂ MW: 194.2464 g/mol |
MDL Number:
MFCD00066453 CAS Number: 1002-62-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...