Dibenzyl sulfoxide ≥99.0% (by HPLC)
Supplier: TCI America
D0001-250G
D0001-25G
TCD0001-250GEA
369.72
CAD
TCD0001-250G
TCD0001-025G
Dibenzyl sulfoxide ≥99.0% (by HPLC)
Dibenzyl sulphoxide
CAS Number: 621-08-9
MDL Number: MFCD00004782
Molecular Formula: C14H14OS
Molecular Weight: 230.33
Purity/Analysis Method: >99.0% (HPLC)
Form: Crystal
Melting point (°C): 133
Product Categories: Materials Chemistry, Pressure & Heat Sensitive Recording Materials
Other Product Names / Synonyms: Benzyl Sulfoxide
Formula:
C₁₄H₁₄OS Melting Pt: 133 °C |
MDL Number:
MFCD00004782 CAS Number: 621-08-9 EINECS: 210-668-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...