Chitosan, 20-100 mPa.s (20 °C) 0,5% in acetic acid (0,5%)
Supplier: TCI America
Synonyms:
Deacetylated chitin, Poly(D-glucosamine)
C2396-25G
C2396-100G
TCC2396-25GEA
62.11
CAD
TCC2396-25G
TCC2396-100G
Chitosan, 20-100 mPa.s (20 °C) 0,5% in acetic acid (0,5%)
Chitosan
CAS Number: 9012-76-4
MDL Number: MFCD00161512
Form: Crystal
Color: Very Pale Yellow
Product Categories: Materials Chemistry, Polymers, Bioscience, Polysaccharides, Natural Polymers (for DDS)
MDL Number:
MFCD00161512 CAS Number: 9012-76-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...