Chitosan, 5-20 mPa.s (20 °C) 0.5% in acetic acid (0,5%)
Supplier: TCI America
C2395-25G
C2395-500G
C2395-100G
TCC2395-25GEA
92.97
CAD
TCC2395-25G
TCC2395-500G
TCC2395-100G
Chitosan, 5-20 mPa.s (20 °C) 0.5% in acetic acid (0,5%)
Chitosan
CAS Number: 9012-76-4
MDL Number: MFCD00161512
Form: Crystal
Color: Slightly Pale Yellow
Product Categories: Materials Chemistry, Polymers, Bioscience, Polysaccharides, Natural Polymers (for DDS)
MDL Number:
MFCD00161512 CAS Number: 9012-76-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...