6-Chloro-3-hydroxypyridazine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 6-chloropyridazin-3-ol, 6-Chloro-3(2H)-pyridazinone

C2377-5G C2377-25G
TCC2377-5GEA 77.42 CAD
TCC2377-5G TCC2377-25G
6-Chloro-3-hydroxypyridazine ≥98.0% (by GC, titration analysis)
6-Chloro-3-hydroxypyridazine

CAS Number: 19064-67-6
MDL Number: MFCD00233955
Molecular Formula: C4H3ClN2O
Molecular Weight: 130.53
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 142


Other Product Names / Synonyms: 3-Chloro-6-hydroxypyridazine

Formula: C₄H₃ClN₂O
MW: 130.53 g/mol
Melting Pt: 142 °C
MDL Number: MFCD00160460
CAS Number: 19064-67-6

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR