Chloro[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene]silver ≥97.0% (by total nitrogen basis)
Supplier: TCI America
Synonyms:
(1,3-Bis(2,6-diisopropylphenyl)-1H-imidazol-2(3H)-ylidene)silver(III) chloride
C2373-1G
C2373-200MG
TCC2373-1GEA
186.44
CAD
TCC2373-1G
TCC2373-200MG
Chloro[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene]silver ≥97.0% (by total nitrogen basis)
Chloro[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene]silver
CAS Number: 873297-19-9
Molecular Formula: C27H36AgClN2
Molecular Weight: 531.92
Purity/Analysis Method: >97.0% (N)
Form: Crystal
Formula:
C₂₇H₃₆AgClN₂ MW: 531.92 g/mol |
MDL Number:
MFCD15072152 CAS Number: 873297-19-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...