6-Chloropyrimidin-4(1H)-one ≥98.0% (by GC, titration analysis)

Supplier: TCI America

C2368-1G C2368-5G
TCC2368-1GEA 55.3 CAD
TCC2368-1G TCC2368-5G
6-Chloropyrimidin-4(1H)-one ≥98.0% (by GC, titration analysis)
6-Chloropyrimidin-4(1H)-one

CAS Number: 4765-77-9
MDL Number: MFCD00233957
Molecular Formula: C4H3ClN2O
Molecular Weight: 130.53
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 195


Other Product Names / Synonyms: 6-Chloro-4-pyrimidinol, 6-Chloro-4(1H)-pyrimidinone

Formula: C₄H₃ClN₂O
Melting Pt: 195 °C
MDL Number: MFCD00233957
CAS Number: 4765-77-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=20244e8bb473b23ffc1882a2a52a61c86958082f&vid=20244e8bb473b23ffc1882a2a52a61c86958082f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR