Cyromazine ≥98.0%

Supplier: TCI America

Synonyms: 2,4-Diamino-6-(cyclopropylamino)-1,3,5-triazine, N2-Cyclopropylmelamine

C2366-5G C2366-25G
TCC2366-5GEA 58.46 CAD
TCC2366-5G TCC2366-25G
Cyromazine ≥98.0%
Cyromazine

CAS Number: 66215-27-8
MDL Number: MFCD00078650
Molecular Formula: C6H10N6
Molecular Weight: 166.19
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 224
Flash Point (°C): 100


Product Categories: Bioscience, Pesticide Ingredients
Other Product Names / Synonyms: 2,4-Diamino-6-(cyclopropylamino)-1,3,5-triazine, N2-Cyclopropylmelamine

Formula: C₆H₁₀N₆
MW: 166.19 g/mol
Melting Pt: 224 °C
Flash Pt: 100 °C
MDL Number: MFCD00078650
CAS Number: 66215-27-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=745aea5b161bcb6f4c6181f69e62ca0dbcb6c6be&vid=745aea5b161bcb6f4c6181f69e62ca0dbcb6c6be&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR