6-Aminofluorescein (isomer II) ≥95.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Fluoresceinamine isomer II, Fluoresceinamine, 6-Amino-3',6'-dihydroxyspiro[isobenzofuran-1(3H),9'-(9H)xanthen]-3-one, 6-Amino-3',6'-dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one, 6-Aminofluorescein, 6-Aminofluorescein (isomer II), 6-Amino-3',6'-dihydroxyspiro[isobenzofuran-1(3H),9'-(9H)xanthen]-3-one (isomer II)
CAS Number: 51649-83-3
MDL Number: MFCD00005051
Molecular Formula: C20H13NO5
Molecular Weight: 347.33
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Melting point (°C): 285
Product Categories: Bioscience, Fluorometry & Fluorescent Imaging, Fluorescent Labeling Dyes, Analytical Science, Fluorescent Dyes
Other Product Names / Synonyms: 6-Amino-3',6'-dihydroxyspiro[isobenzofuran-1(3H),9'-(9H)xanthen]-3-one, Fluoresceinamine
Formula:
C₂₀H₁₃NO₅ MW: 347.33 g/mol Melting Pt: 285 °C |
MDL Number:
MFCD00005051 CAS Number: 51649-83-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...