2-Chloro-4-hydroxybenzoic acid 98,0
Supplier: TCI America
C2349-1G
C2349-5G
TCC2349-1GEA
213.3
CAD
TCC2349-1G
TCC2349-5G
2-Chloro-4-hydroxybenzoic acid 98,0
2-Chloro-4-hydroxybenzoic acid
CAS Number: 56363-84-9
MDL Number: MFCD00045855
Molecular Formula: C7H5ClO3
Molecular Weight: 172.56
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 159
Formula:
C₇H₅ClO₃ MW: 172.57 Boiling Pt: 350 Melting Pt: 159 °C |
MDL Number:
MFCD00045855 CAS Number: 56363-84-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...