7-Chloro-6-nitro-4-hydroxyquinazoline ≥98.0%
Supplier: TCI America
Synonyms:
7-Chloro-6-nitroquinazolin-4(3H)-one, 7-Chloro-6-nitro-4(3H)-quinazolinone, 7-Chloro-6-nitro-4-quinazolinol, 7-Chloro-6-nitro-4-quinazolinone
C2332-25G
C2332-5G
TCC2332-25GEA
297.04
CAD
TCC2332-25G
TCC2332-5G
7-Chloro-6-nitro-4-hydroxyquinazoline ≥98.0%
7-Chloro-6-nitro-4-hydroxyquinazoline
CAS Number: 53449-14-2
MDL Number: MFCD07772904
Molecular Formula: C8H4ClN3O3
Molecular Weight: 225.59
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 303
Other Product Names / Synonyms: 7-Chloro-6-nitro-4-quinazolinol, 7-Chloro-6-nitro-4-quinazolinone
Formula:
C₈H₄ClN₃O₃ MW: 225.59 g/mol Melting Pt: 303 °C |
MDL Number:
MFCD09388771 CAS Number: 53449-14-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...