1,1'-Carbonyldi(1H-1,2,4-triazole) ≥97.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Di(1H-1,2,4-triazol-1-yl)methanone
C2325-5G
C2325-25G
TCC2325-5GEA
238.58
CAD
TCC2325-5G
TCC2325-25G
1,1'-Carbonyldi(1H-1,2,4-triazole) ≥97.0% (by titrimetric analysis)
1,1'-Carbonyldi(1H-1,2,4-triazole)
CAS Number: 41864-22-6
MDL Number: MFCD00043399
Molecular Formula: C5H4N6O
Molecular Weight: 164.13
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 150
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Nucleic Acid Synthesis
Other Product Names / Synonyms: 1,1'-Carbonylbis(1,2,4-triazole), CDT
Formula:
C₅H₄N₆O MW: 164.1249 g/mol Melting Pt: 150 °C |
MDL Number:
MFCD00043399 CAS Number: 41864-22-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...