Acetrizoic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI America
A0858-1G
A0858-5G
TCA0858-001GEA
83.74
CAD
TCA0858-001G
TCA0858-5G
Acetrizoic acid ≥98.0% (by titrimetric analysis)
Acetrizoic acid
CAS Number: 85-36-9
MDL Number: MFCD00016495
Molecular Formula: C9H6I3NO3
Molecular Weight: 556.86
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Other Product Names / Synonyms: 3-Acetamido-2,4,6-triiodobenzoic Acid
Formula:
C₉H₆I₃NO₃ |
MDL Number:
MFCD00016495 CAS Number: 85-36-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...