trans-1,4-Cyclohexanediol ≥97.0%

Supplier: TCI America

Synonyms: trans-Cyclohexane-1,4-diol, trans-1,4-Dihydroxycyclohexane

C2320-1G C2320-5G
TCC2320-1GEA 406.69 CAD
TCC2320-1G TCC2320-5G
trans-1,4-Cyclohexanediol ≥97.0%
trans-1,4-Cyclohexanediol

CAS Number: 6995-79-5
MDL Number: MFCD00063612
Molecular Formula: C6H12O2
Molecular Weight: 116.16
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 140


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: trans-1,4-Dihydroxycyclohexane

Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Melting Pt: 140 °C
MDL Number: MFCD00063612
CAS Number: 6995-79-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0919bc9d10a52895828a90093ac860dd9cefec37&vid=0919bc9d10a52895828a90093ac860dd9cefec37&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR