cis-1,2-Cyclohexanedimethanol ≥97.0%

Supplier: TCI America

C2318-1G C2318-5G
TCC2318-1GEA 101.12 CAD
TCC2318-1G TCC2318-5G
cis-1,2-Cyclohexanedimethanol ≥97.0%
cis-1,2-Cyclohexanedimethanol

CAS Number: 15753-50-1
MDL Number: MFCD00062987
Molecular Formula: C8H16O2
Molecular Weight: 144.21
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 112
Melting point (°C): 40


Other Product Names / Synonyms: cis-1,2-Bis(hydroxymethyl)cyclohexane

Formula: C₈H₁₆O₂
Boiling Pt: 112 °C
Melting Pt: 40 °C
MDL Number: MFCD00062987
CAS Number: 15753-50-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c95cedd79c6c835f8776a8b3dc944871db65ec4b&vid=c95cedd79c6c835f8776a8b3dc944871db65ec4b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR