Chloro[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene]copper(I) ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Chloro[1,3-bis(2,6-di-i-propylphenyl)imidazol-2-ylidene]copper(I), [1,3-Bis(2,6-diisopropylphenyl)imidazol-2-ylidene]copper chloride
C2304-1G
C2304-200MG
TCC2304-1GEA
254.38
CAD
TCC2304-1G
TCC2304-200MG
Chloro[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene]copper(I) ≥98.0% (by titrimetric analysis)
Chloro[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene]copper(I)
CAS Number: 578743-87-0
MDL Number: MFCD09264276
Molecular Formula: C27H36ClCuN2
Molecular Weight: 487.60
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Formula:
C₂₇H₃₆ClCuN₂ MW: 487.60 g/mol |
MDL Number:
MFCD09264276 CAS Number: 578743-87-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...