4-Aminostilbene ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America

A0854-25G A0854-5G
TCA0854-25GEA 2509.04 CAD
TCA0854-25G TCA0854-005G
4-Aminostilbene ≥97.0% (by HPLC, titration analysis)
4-Aminostilbene

CAS Number: 834-24-2
MDL Number: MFCD00025377
Molecular Formula: C14H13N
Molecular Weight: 195.27
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal
Melting point (°C): 151


Other Product Names / Synonyms: 4-Stilbenamine

Formula: C₁₄H₁₃N
Melting Pt: 151 °C
MDL Number: MFCD00025377
CAS Number: 834-24-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1e4d938d7f479898973d1ea9de7f93d98a499346&vid=1e4d938d7f479898973d1ea9de7f93d98a499346&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR