7-Chloro-1-cyclopropyl-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
C2293-5G
C2293-25G
TCC2293-5GEA
56.88
CAD
TCC2293-5G
TCC2293-25G
7-Chloro-1-cyclopropyl-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid ≥98.0% (by HPLC, titration analysis)
7-Chloro-1-cyclopropyl-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid
CAS Number: 86393-33-1
MDL Number: MFCD00792458
Molecular Formula: C13H9ClFO3N
Molecular Weight: 281.67
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 244
Product Categories: Synthetic Organic Chemistry, Fluorinated Bioactive Compounds, Bioscience, Synthetic Antimicrobial Ingredients
Other Product Names / Synonyms: Fluoroquinolonic Acid
Formula:
C₁₃H₉ClFNO₃ Melting Pt: 244 °C |
MDL Number:
MFCD00792458 CAS Number: 86393-33-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...