4-Chloro-1-naphthol ≥98.0% (by titrimetric analysis) for biochemical research

Supplier: TCI America

C2291-5G C2291-1G
TCC2291-5GEA 115.34 CAD
TCC2291-5G TCC2291-1G
4-Chloro-1-naphthol ≥98.0% (by titrimetric analysis) for biochemical research
4-Chloro-1-naphthol

[for Biochemical Research]
CAS Number: 604-44-4
MDL Number: MFCD00003974
Molecular Formula: C10H7ClO
Molecular Weight: 178.62
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 120


Product Categories: Bioscience, Stains & Dyes, Peroxidase Substrates
Other Product Names / Synonyms: 4-CN

Formula: ClC₁₀H₆OH
Melting Pt: 120 °C
MDL Number: MFCD00003974
CAS Number: 604-44-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=64cef96f043f6bcf6d00ed37fc8cce44eb92e9f0&vid=151a6dff8e0a0fa58a3dabbbdb1c2d3fbaffeede&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR