(1ɑ,3ɑ,5ɑ)-1,3,5-Cyclohexanetriol ≥98.0%

Supplier: TCI America

Synonyms: (1α,3α,5α)-1,3,5-Cyclohexanetriol, cis,cis-1,3,5-Trihydroxycyclohexane, cis-Phloroglucitol

C2275-5G C2275-25G
TCC2275-5GEA 432.92 CAD
TCC2275-5G TCC2275-25G
(1ɑ,3ɑ,5ɑ)-1,3,5-Cyclohexanetriol ≥98.0%
(1alpha,3alpha,5alpha)-1,3,5-Cyclohexanetriol

CAS Number: 50409-12-6
MDL Number: MFCD00043062
Molecular Formula: C6H12O3
Molecular Weight: 132.16
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 188

Formula: C₆H₁₂O₃
MW: 132.16 g/mol
Melting Pt: 188 °C
MDL Number: MFCD00043062
CAS Number: 50409-12-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8139a34967ff75f5d62c69b80970c11d2886c5b1&vid=8139a34967ff75f5d62c69b80970c11d2886c5b1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR