1,2,3,4-Cyclobutanetetracarboxylic dianhydride ≥98.0%

Supplier: TCI America

Synonyms: Cyclobuta[1,2-c:3,4-c']difuran-1,3,4,6(3aH,3bH,6aH,6bH)-tetraone

C2262-1G C2262-5G
TCC2262-1GEA 96.38 CAD
TCC2262-1G TCC2262-5G
1,2,3,4-Cyclobutanetetracarboxylic dianhydride ≥98.0%
1,2,3,4-Cyclobutanetetracarboxylic dianhydride

CAS Number: 4415-87-6
MDL Number: MFCD00004944
Molecular Formula: C8H4O6
Molecular Weight: 196.11
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 285

Formula: C₈H₄O₆
MW: 196.11 g/mol
Melting Pt: 285 °C
MDL Number: MFCD00004944
CAS Number: 4415-87-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=45dfa05f3063eb763a128906034a6253013c8aff&vid=45dfa05f3063eb763a128906034a6253013c8aff&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR