N-Acetyl-DL-2-aminobutyric acid 97,0

Supplier: TCI America

Synonyms: 2-Acetamidobutyric Acid

A0853-25G A0853-5G
TCA0853-025GEA 586.81 CAD
TCA0853-025G TCA0853-5G
N-Acetyl-DL-2-aminobutyric acid 97,0
N-Acetyl-DL-2-aminobutyric acid

CAS Number: 7211-57-6
MDL Number: MFCD00020430
Molecular Formula: C6H11NO3
Molecular Weight: 145.16
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 132


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: 2-Acetamidobutyric Acid

Formula: C₆H₁₁NO₃
MW: 145.16 g/mol
Melting Pt: 132 °C
MDL Number: MFCD00020430
CAS Number: 7211-57-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b86c62ea18184a0a1ccc01d2a653609aaceb7531&vid=b86c62ea18184a0a1ccc01d2a653609aaceb7531&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR