Chlorobis(cyclooctene)rhodium(I) dimer ≥96.0% (by titrimetric analysis)

Supplier: TCI America

C2253-100MG C2253-500MG
TCC2253-100MGEA 178.54 CAD
TCC2253-100MG TCC2253-500MG
Chlorobis(cyclooctene)rhodium(I) dimer ≥96.0% (by titrimetric analysis)
Chlorobis(cyclooctene)rhodium(I) dimer

CAS Number: 12279-09-3
MDL Number: MFCD00013287
Molecular Formula: C32H56Cl2Rh2
Molecular Weight: 717.51
Purity/Analysis Method: >96.0% (T)
Form: Crystal
Melting point (°C): 190
Storage Temperature: <0°C

Formula: C₃₂H₅₆Cl₂Rh₂
MW: 717.51 g/mol
Melting Pt: 190 °C
MDL Number: MFCD00013287
CAS Number: 12279-09-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3e482a09376f0d4e05089509c3b32706275eb078&vid=3e482a09376f0d4e05089509c3b32706275eb078&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR