5-Chloro-3-nitrosalicylaldehyde ≥98.0% (by GC, titration analysis)
Supplier: TCI America
C2249-1G
C2249-5G
TCC2249-1GEA
97.96
CAD
TCC2249-1G
TCC2249-5G
5-Chloro-3-nitrosalicylaldehyde ≥98.0% (by GC, titration analysis)
5-Chloro-3-nitrosalicylaldehyde
CAS Number: 16634-90-5
MDL Number: MFCD00462959
Molecular Formula: C7H4ClNO4
Molecular Weight: 201.56
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 110
Other Product Names / Synonyms: 5-Chloro-2-hydroxy-3-nitrobenzaldehyde
Formula:
C₇H₄ClNO₄ Melting Pt: 110 °C |
MDL Number:
MFCD00462959 CAS Number: 16634-90-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...