Gimeracil ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 5-Chloro-4-hydroxypyridin-2(1H)-one, 5-Chloro-4-hydroxy-2(1H)-pyridinone, 5-Chloro-2,4-pyridinediol, 5-Chloro-2,4-dihydroxypyridine

C2243-1G C2243-5G
TCC2243-1GEA 143.04 CAD
TCC2243-1G TCC2243-5G
Gimeracil ≥98.0% (by HPLC, titration analysis)
Gimeracil

CAS Number: 103766-25-2
MDL Number: MFCD08458352
Molecular Formula: C5H4ClNO2
Molecular Weight: 145.54
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 274


Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Antitumor Ingredients
Other Product Names / Synonyms: 5-Chloro-2,4-dihydroxypyridine, 5-Chloro-2,4-pyridinediol, 5-Chloro-4-hydroxy-2(1H)-pyridinone

Formula: C₅H₄ClNO₂
MW: 145.54 g/mol
Melting Pt: 274 °C
MDL Number: MFCD08458352
CAS Number: 103766-25-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e4cb5e6a0adf1b64c3eedb32705763d9ccd8441f&vid=e4cb5e6a0adf1b64c3eedb32705763d9ccd8441f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR